N,N-Dimethylacetamid dimethyl acetal ≥90.0% (by titrimetric analysis) stabilised
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
(1,1-Dimethoxyethyl)dimethylamine, 1,1-Dimethoxy-N,N-dimethylethanamine
Stabilization:
stabiliseret med methanol 5-10%
D1652-25ML
D1652-100ML
TCIAD1652-25MLEA
887
DKK
TCIAD1652-25ML
TCIAD1652-100ML
N,N-Dimethylacetamid dimethyl acetal ≥90.0% (by titrimetric analysis) stabilised
N,N-Dimethylacetamid dimethyl acetal
Formel:
C₆H₁₅NO₂ Flammepunkt: 8°C |
MDL Number:
MFCD00008476 CAS-nummer: 18871-66-4 EINECS: 242-641-0 UN nummer: 1993 ADR: 3,II |
|
Specifikationer Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (Nonaqueous Titration) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...