4-Nitro-o-xylen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3,4-Dimethyl-1-nitrobenzene, 3,4-Dimethylnitrobenzene, 1,2-Dimethyl-4-nitrobenzene, 4-Nitro-o-xylene

D1085-25G D1085-500G
TCIAD1085-25GEA 132 DKK
TCIAD1085-25G TCIAD1085-500G
4-Nitro-o-xylen ≥98.0% (by GC)
4-Nitro-o-xylen
Formel: C₈H₉NO₂
Kogepunkt: 254 °C (1013 hPa)
Smeltepunkt: 31…32 °C
Massefylde: 1,117 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007268
CAS-nummer: 99-51-4
EINECS: 202-761-6
UN nummer: 3447
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34d3e0f42612e3772b6ed8bb3e98dd7a04f899&vid=1b41c544b342186369f997848634e8576145e117&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR