4-(Benzyloxy)benzoesyre ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B1665-25G B1665-5G
TCIAB1665-25GEA 952 DKK
TCIAB1665-25G TCIAB1665-5G
4-(Benzyloxy)benzoesyre ≥98.0% (by GC, titration analysis)
4-(Benzyloxy)benzoesyre
Formel: C₁₄H₁₂O₃
Molekylvægt: 228.25 g/mol
MDL Number: MFCD00016527
CAS-nummer: 1486-51-7
EINECS: 216-066-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7853af38e84099bf8247390ce0d0a2491b34be98&vid=7853af38e84099bf8247390ce0d0a2491b34be98&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 189.0 to 193.0 deg-C
Solubility in Pyridine very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR