(±)-Methyl 2-methylbutyrat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Methyl2-methylbutyrate

M0758-25ML M0758-100ML
TCIAM0758-25MLEA 148 DKK
TCIAM0758-25ML TCIAM0758-100ML
(±)-Methyl 2-methylbutyrat ≥98.0% (by GC)
(±)-Methyl 2-methylbutyrat
Formel: C₆H₁₂O₂
Molekylvægt: 116.16 g/mol
Kogepunkt: 115°C
Flammepunkt: 21°C
MDL Number: MFCD00009335
CAS-nummer: 868-57-5
EINECS: 212-778-0
UN nummer: 3272
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2982faff22754ca99563bc8df0f471e920abae2&vid=b2982faff22754ca99563bc8df0f471e920abae2&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR