1H,1H,2H-Perfluor-1-octen ≥96.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

P1102-25G P1102-5G P1102-100G
TCIAP1102-25GEA 591 DKK
TCIAP1102-25G TCIAP1102-5G TCIAP1102-100G
1H,1H,2H-Perfluor-1-octen ≥96.0% (by GC)
1H,1H,2H-Perfluor-1-octen
Formel: C₈H₃F₁₃
Kogepunkt: 103°C
Massefylde: 1,56 g/cm³ (20 °C)
MDL Number: MFCD00039249
CAS-nummer: 25291-17-2
EINECS: 246-791-8
UN nummer: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e28ee71d4a5ba1df4da0ded3e0bd88798ae5100&vid=b3fd11add6b2737ae0dbe4030a375a59f35bbe2e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR