Bromacetaldehyddimethylacetal ≥97.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Bromo-1,1-dimethoxyethane

Stabilization: Stabilised with potassium carbonate

B1097-25G B1097-500G B1097-100G
TCIAB1097-25GEA 140 DKK
TCIAB1097-25G TCIAB1097-500G TCIAB1097-100G
Bromacetaldehyddimethylacetal ≥97.0% (by GC) stabilised
Bromacetaldehyddimethylacetal
Formel: BrH₂CCH(OCH₃)₂
Molekylvægt: 169.02 g/mol
Kogepunkt: 150°C
Flammepunkt: 52°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00000213
CAS-nummer: 7252-83-7
EINECS: 230-669-6
UN nummer: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b0eb268128909ffa6b84979fe04a8833bf4ab5a&vid=1fc07f1e7141552e9eda6fc27fababb840096266&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR