Borane dimethyl sulphide complex ≥90.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Borane-methyl sulphide complex
D1843-25ML
D1843-100ML
TCIAD1843-25MLEA
255
DKK
TCIAD1843-25ML
TCIAD1843-100ML
Borane dimethyl sulphide complex ≥90.0% (by titrimetric analysis)
Borane dimethyl sulphide complex
Formel:
C₂H₉BS Smeltepunkt: –27 °C Massefylde: 1,28 g/cm³ (20 °C) Flammepunkt: 18°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00013189 CAS-nummer: 13292-87-0 EINECS: 236-313-6 UN nummer: 3399 ADR: 4.3,I |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (Neutralization titration) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...