2,3-Butandiol mixture of stereoisomers ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
2,3-Dihydroxy butane, (±)-2,3-Butanediol, 2,3-Butylene glykol
B0681-25G
B0681-500G
B0681-100G
TCIAB0681-25GEA
156
DKK
TCIAB0681-25G
TCIAB0681-500G
TCIAB0681-100G
2,3-Butandiol mixture of stereoisomers ≥97.0% (by GC)
2,3-Butanediol
Formel:
C₄H₁₀O₂ Kogepunkt: 182°C Smeltepunkt: 18…20 °C Massefylde: 1 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004523 CAS-nummer: 513-85-9 EINECS: 208-173-6 Merck Index: 14,01568 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % (total of diastereoisomers) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...