1-Chlor-2-iodoethan ≥97.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel


Stabilization: Stabilised with copper chip

C1190-1G C1190-5G
TCIAC1190-1GEA 1010 DKK
TCIAC1190-1G TCIAC1190-5G
1-Chlor-2-iodoethan ≥97.0% (by GC) stabilised
1-Chlor-2-iodoethan
Formel: C₂H₄ClI
Molekylvægt: 190.41 g/mol
Kogepunkt: 140°C
Smeltepunkt: -15.6
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00041453
CAS-nummer: 624-70-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d808b6e605746013a7f3c4ecc3ff670582ed6df6&vid=d808b6e605746013a7f3c4ecc3ff670582ed6df6&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR