D(+)-Xylose ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
X0019-500G
X0019-25G
TCIAX0019-500GEA
641
DKK
TCIAX0019-500G
TCIAX0019-25G
D(+)-Xylose ≥98.0% (by HPLC)
D(+)-Xylose
Formel:
C₅H₁₀O₅ Molekylvægt: 150.13 g/mol Kogepunkt: 331 Smeltepunkt: 154 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00151475 CAS-nummer: 58-86-6 EINECS: 200-400-7 Merck Index: 14,10087 |
|
Specifikationer Testresultater
Appearance | White powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 146.0 to 153.0 deg-C |
Specific rotation [a]20/D | +19.0 to +21.0 deg (C=10, H2O) |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...