Octan-1-sulfonsyre natrium salt ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Sodium octane-1-sulphonate
O0123-25G
O0123-500G
O0123-100G
TCIAO0123-25GEA
550
DKK
TCIAO0123-25G
TCIAO0123-500G
TCIAO0123-100G
Octan-1-sulfonsyre natrium salt ≥98.0% (by titrimetric analysis)
Octan-1-sulfonsyre natrium salt
Formel:
C₈H₁₇NaO₃S Molekylvægt: 216.28 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007544 CAS-nummer: 5324-84-5 EINECS: 226-195-4 |
|
Specifikationer Testresultater
Appearance | White powder to crystal |
Purity (Ion exchange titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...