Glutaraldehyd 24 - 26% in water

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Glutaral

G0067-500ML G0067-25ML
TCIAG0067-500MLEA 2510 DKK
TCIAG0067-500ML TCIAG0067-25ML
Glutaraldehyd 24 - 26% in water
Glutaraldehyd
Formel: C₅H₈O₂
Molekylvægt: 100.12 g/mol
Kogepunkt: 188
Smeltepunkt: -10
Massefylde: 1.014
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00007025
CAS-nummer: 111-30-8
EINECS: 203-856-5
UN nummer: 2927
ADR: 6.1 II
Merck Index: 14,04472
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c2fbca3c625484220b2ce95b83ea86a8a75bf01&vid=e706d354dac2766116cc4d05eedf7783d53af3a5&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless clear liquid
Concentration (Oxime Formation) 24.0 to 26.0 w/w%
abs-ratio (a/b) max. 0.35 (E230/280)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR