trans-2-Phenylcyclohexanol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: trans-2-Phenyl-1-cyclohexanol

P1174-1G P1174-5G
TCIAP1174-1GEA 591 DKK
TCIAP1174-1G TCIAP1174-5G
trans-2-Phenylcyclohexanol ≥98.0% (by GC)
trans-2-Phenylcyclohexanol
Formel: C₁₂H₁₆O
MDL Number: MFCD00003865
CAS-nummer: 2362-61-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98ed1228856f8f3d3690d0e98244fbd473a1dcfc&vid=98ed1228856f8f3d3690d0e98244fbd473a1dcfc&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 56.0 to 60.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR