Diallylsulfid ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Allyl sulphide
A0235-25ML
A0235-250ML
TCIAA0235-25MLEA
214
DKK
TCIAA0235-25ML
TCIAA0235-250ML
Diallylsulfid ≥98.0% (by GC)
Diallylsulfid
Formel:
C₆H₁₀S Molekylvægt: 114.21 g/mol Kogepunkt: 138°C Smeltepunkt: -83 Flammepunkt: 46°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008658 CAS-nummer: 592-88-1 EINECS: 209-775-1 UN nummer: 1993 ADR: 3 III Merck Index: 14,00297 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...