D(-)-Vinsyre 99,0 Titrimetric
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
unnatural tartaric acid, (2S,3S)-2,3-Dihydroxysuccinic acid, (S,S)-(-)-Tartaric acid, (S,S)-(-)-2,3-Dihydroxysuccinic acid
T0026-500G
T0026-25G
T0026-100G
TCIAT0026-500GEA
2020
DKK
TCIAT0026-500G
TCIAT0026-25G
TCIAT0026-100G
D(-)-Vinsyre 99,0 Titrimetric
D(-)-Vinsyre
Formel:
C₄H₆O₆ Molekylvægt: 150.09 g/mol Kogepunkt: 356 Flammepunkt: 210 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004238 CAS-nummer: 147-71-7 EINECS: 205-695-6 Merck Index: 14,09068 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 99.0 % |
Specific rotation [a]20/D | -11.5 to -13.0 deg (C=20, H2O) |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...