1-Hydroxy-9,10-anthraquinone ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Hydroxyanthraquinone
H0354-25G
H0354-5G
TCIAH0354-25GEA
977
DKK
TCIAH0354-25G
TCIAH0354-5G
1-Hydroxy-9,10-anthraquinone ≥98.0% (by HPLC)
1-Hydroxy-9,10-anthraquinone
Formel:
C₁₄H₈O₃ Molekylvægt: 224.2115 g/mol |
MDL Number:
MFCD00058946 CAS-nummer: 129-43-1 EINECS: 204-946-7 |
|
Specifikationer Testresultater
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 195.0 to 199.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...