Zirconium disulphate tetrahydrate ≥98% (metals basis)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
41041.22
41041.36
41041.A3
41041.22EA
601
DKK
41041.22
41041.36
41041.A3
Zirconium disulphate tetrahydrate ≥98% (metals basis)
Zirconium disulphate tetrahydrate
Purity: 98+%
CAS No.: 7446-31-3
Molecular Formula: Zr(SO4)2.4H2O
Formula Weight: 355.40 (283.34anhy)
Physical Form: -20 Mesh
Density: 3.22
Appearance: White
Solubility: Soluble in water
Merck Reference: 14,10182
MDL No.: MFCD00149267
Catalyst support
Formel:
O₈S₂Zr·4H₂O Massefylde: 3,22 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00149267 CAS-nummer: 7446-31-3 EINECS: 238-694-4 Merck Index: 14,10182 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...