Indium(III) hydroxide ≥99.998% (metals basis)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
11855.09
11855.18
11855.09EA
1250
DKK
11855.09
11855.18
Indium(III) hydroxide ≥99.998% (metals basis)
Indium(III) hydroxide
Purity: 99.998%
CAS No.: 20661-21-6
Molecular Formula: In(OH)3
Formula Weight: 165.84
Physical Form: -100 Mesh Powder
Melting Point: 150° -H2O
MDL No.: MFCD00011056
Formel:
H₃InO₃ Molekylvægt: 165.84 g/mol Smeltepunkt: 150 °C (-H₂O) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011056 CAS-nummer: 20661-21-6 EINECS: 243-947-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...