Methyl-4-fluoro-3-hydroxybenzoate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methyl 4-fluoro-3-hydroxybenzoate

M1919-25G M1919-5G
TCIAM1919-25GEA 2290 DKK
TCIAM1919-25G TCIAM1919-5G
Methyl-4-fluoro-3-hydroxybenzoate ≥98.0% (by GC)
Methyl-4-fluoro-3-hydroxybenzoate
Formel: C₈H₇FO₃
Molekylvægt: 170.14 g/mol
MDL Number: MFCD06797457
CAS-nummer: 214822-96-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22f444740de95fa0ac74fcee8878c1aac15bc845&vid=22f444740de95fa0ac74fcee8878c1aac15bc845&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 90.0 to 94.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR