Methyl-4-fluoro-3-hydroxybenzoate ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Methyl 4-fluoro-3-hydroxybenzoate
B25683.14
B25683.06
B25683.14EA
2680
DKK
B25683.14
B25683.06
Methyl-4-fluoro-3-hydroxybenzoate ≥98%
Methyl-4-fluoro-3-hydroxybenzoate
Purity: 98+%
CAS No.: 214822-96-5
Molecular Formula: C8H7FO3
Formula Weight: 170.14
Melting Point: 89-91°
MDL No.: MFCD06797457
Formel:
C₈H₇FO₃ Molekylvægt: 170.14 g/mol Smeltepunkt: 89…91 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD06797457 CAS-nummer: 214822-96-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...