(±)-Aspartic acid ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-Aminobutanedioic acid, DL-Aspartic acid

A0544-25G A0544-500G
TCIAA0544-25GEA 132 DKK
TCIAA0544-25G TCIAA0544-500G
(±)-Aspartic acid ≥98.0% (by titrimetric analysis)
(±)-Aspartic acid
Formel: C₄H₇NO₄
Smeltepunkt: 280°C
MDL Number: MFCD00063083
CAS-nummer: 617-45-8
EINECS: 210-513-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db43af7b2b1a1e16a9aa458b67204344fb535c0d&vid=115d75703194ca8611108689871c6d34dc527549&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in HCl almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR