Benzyl methyl ether ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(Methoxymethyl)benzene
B0424-25ML
B0424-500ML
TCIAB0424-25MLEA
386
DKK
TCIAB0424-25ML
TCIAB0424-500ML
Benzyl methyl ether ≥97.0% (by GC)
Benzyl methyl ether
Formel:
C₈H₁₀O Flammepunkt: 56°C |
MDL Number:
MFCD00025901 CAS-nummer: 538-86-3 EINECS: 208-705-7 UN nummer: 3271 ADR: 3,III Merck Index: 14,01140 |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...