Benzyl chloromethyl ether ≥90.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
((Chloromethoxy)methyl)benzene
B1207.0500
B1207-25G
B1207-500G
B1207-100G
TCIAB1207.0500EA
0
DKK
TCIAB1207.0500
TCIAB1207-25G
TCIAB1207-500G
TCIAB1207-100G
Benzyl chloromethyl ether ≥90.0% (by GC)
Benzyl chloromethyl ether
Formel:
C₈H₉ClO Kogepunkt: 56°C Flammepunkt: 91°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00000886 CAS-nummer: 3587-60-8 UN nummer: 2810 ADR: 6.1,III |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...