4-Bromo-2,3,5,6-tetrafluorobenzoic acid ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H32982.03 H32982.06
H32982.03EA 1050 DKK
H32982.03 H32982.06
4-Bromo-2,3,5,6-tetrafluorobenzoic acid ≥97%
4-Bromo-2,3,5,6-tetrafluorobenzoic acid

Purity: 97+% CAS No.: 4707-24-8 Molecular Formula: C7HBrF4O2 Formula Weight: 272.98 Odour: Odourless Melting Point: 142-145° MDL No.: MFCD00075244

Formel: C₇HBrF₄O₂
Smeltepunkt: 142…145 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00075244
CAS-nummer: 4707-24-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR