N,N'-Diisopropylethylenediamine 97%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 97%
CAS No.: 4013-94-9
Molecular Formula: C8H20N2
Formula Weight: 144.26
Flash Point: 51°(123°F)
Refractive Index: 1.4289
Boiling Point: 169-171°
Beilstein Reference: 1735571
UN No.: 2733
MDL No.: MFCD00008864
Formel:
C₈H₂₀N₂ Kogepunkt: 169…171 °C Massefylde: 0,798 Flammepunkt: 51 °C (123 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008864 CAS-nummer: 4013-94-9 EINECS: 223-666-6 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...