4,4'-Dichloro-trans-stilbene ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
(E)-1,2-Bis(4-chlorophenyl)ethene
D3063-1G
D3063-250MG
TCIAD3063-1GEA
2080
DKK
TCIAD3063-1G
TCIAD3063-250MG
4,4'-Dichloro-trans-stilbene ≥98.0% (by GC)
4,4'-Dichloro-trans-stilbene
Formel:
C₁₄H₁₀Cl₂ Molekylvægt: 249.135 g/mol Kogepunkt: 230°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD01108802 CAS-nummer: 1657-56-3 |
|
Specifikationer Testresultater
Appearance | White to Gray to Dark blue powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Melting point | 175.0 to 179.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...