2,2'-Bithiophene-5-boronic acid pinacol ester ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

T2518-1G T2518-5G T2518-25G
TCIAT2518-1GEA 271 DKK
TCIAT2518-1G TCIAT2518-5G TCIAT2518-25G
2,2'-Bithiophene-5-boronic acid pinacol ester ≥98.0% (by GC)
2,2'-Bithiophene-5-boronic acid pinacol ester
Formel: C₁₄H₁₇BO₂S₂
MDL Number: MFCD04039974
CAS-nummer: 479719-88-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e796a91cd4eb4f7b63a2788e8e957087aedaba45&vid=e796a91cd4eb4f7b63a2788e8e957087aedaba45&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Green to Blue powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR