Pentatitanium trisilicide ≥99.5% (metals basis)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Titanium silicide

13087.18 13087.30
13087.18EA 1700 DKK
13087.18 13087.30
Pentatitanium trisilicide ≥99.5% (metals basis)
Pentatitanium trisilicide

Purity: 99.5% CAS No.: 12067-57-1 Molecular Formula: Ti5Si3 Formula Weight: 323.76 Physical Form: -325 Mesh Powder Odour: Odourless UN No.: 3178 MDL No.: MFCD01310208

Formel: Si₃Ti₅
Molekylvægt: 323.76 g/mol
Smeltepunkt: 2130 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01310208
CAS-nummer: 12067-57-1
EINECS: 235-082-9
UN nummer: 3178
ADR: 4.1 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR