Sodium-4-aminosalicylate dihydrate ~99%, white powder

Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Sodium 4-Aminosalicylate Dihydrate, 4-Amino-2-hydroxybenzoic acid sodium salt, 4-Aminosalicylic acid sodium salt, Sodium 4-amino-2-hydroxybenzoate dihydrate

0215036790 0215036725
ICNA0215036790EA 823 DKK
ICNA0215036790 ICNA0215036725
Sodium-4-aminosalicylate dihydrate ~99%, white powder
Sodium-4-aminosalicylate dihydrate

-Aminosalicylic acid is an analogue of p-aminobenzoic acid. It is an antitubercular agent. The sodium salt of the drug is better tolerated than the free acid.


Applications
p-Aminosalicylic acid has antibacterial properties. It is used to inhibit growth and multiplication of the tubercle bacillus.


Soluble in water (1 g/2 ml); sparingly soluble in acetone; practically insoluble in ether, chloroform or benzene. Sodium salt solutions is more resistant to heat than the free acid, but sterilisation by filtration is recommended.

Formel: C₇H₆NNaO₃·2H₂O
Smeltepunkt: 150 °C (dec., lit.)
MDL Number: MFCD00064392
CAS-nummer: 6018-19-5
EINECS: 205-091-2

Bestil nu

Specifikationer Testresultater

Identity Test Passes
Purity ~99%
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR