Farnesyl thiosalicylic acid amide

Leverandør: Cayman Chemical
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

13175-1 13175-5 13175-10 13175-50
CAYM13175-1EA 410 DKK
CAYM13175-1 CAYM13175-5 CAYM13175-10 CAYM13175-50
Farnesyl thiosalicylic acid amide
Farnesyl thiosalicylic acid amide
Formel: C₂₂H₃₁NOS
CAS-nummer: 1092521-74-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR