3-(Trimethylsilyl)phenylboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (3-(Trimethylsilyl)phenyl)boronic acid

T2663-5G T2663-1G
TCIAT2663-5GEA 1020 DKK
TCIAT2663-5G TCIAT2663-1G
3-(Trimethylsilyl)phenylboronic acid (contains varying amounts of Anhydride)
3-(Trimethylsilyl)phenylboronic acid
Formel: C₉H₁₅BO₂Si
MDL Number: MFCD03093884
CAS-nummer: 177171-16-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72bb2e84155aadc76914d3d5c5d526906888a9e1&vid=72bb2e84155aadc76914d3d5c5d526906888a9e1&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 113.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR