Potassium allyltrifluoroborate ≥97.0% (W)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

P1480-1G P1480-5G
TCIAP1480-1GEA 444 DKK
TCIAP1480-1G TCIAP1480-5G
Potassium allyltrifluoroborate ≥97.0% (W)
Potassium allyltrifluoroborate
Formel: C₃H₅BF₃K
MDL Number: MFCD07368222
CAS-nummer: 233664-53-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13571ab2e3b5bb67f0b5ba0fb3ff7887c53560a9&vid=13571ab2e3b5bb67f0b5ba0fb3ff7887c53560a9&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Gravimetric) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR