9-Methylcarbazole ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
9-Methyl-9H-carbazole, N-Methylcarbazole
M0960-25G
M0960-5G
TCIAM0960-25GEA
2060
DKK
TCIAM0960-25G
TCIAM0960-5G
9-Methylcarbazole ≥99.0% (by GC)
9-Methylcarbazole
Formel:
C₁₃H₁₁N Kogepunkt: 195°C |
MDL Number:
MFCD00013431 CAS-nummer: 1484-12-4 EINECS: 216-054-5 |
|
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 88.0 to 91.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...