Methyl glycolate 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Hydroxyacetic Acid Methyl Ester, Methyl 2-hydroxyacetate, Glycolic Acid Methyl Ester, Methyl Hydroxyacetate

414370250 414371000
ACRO414370250EA 1220 DKK
ACRO414370250 ACRO414371000
Methyl glycolate 98%
Methyl glycolate
Formel: C₃H₆O₃
Molekylvægt: 90.08 g/mol
Kogepunkt: 52 °C (17 mmHg)
Smeltepunkt: 149…151 °C
Massefylde: 1.184 g/cm³
Flammepunkt: 67 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004667
CAS-nummer: 96-35-5
EINECS: 202-502-7

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4150 to 1.4180 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR