2-Hydroxyethyl acetate >60.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

A1059-25ML A1059-500ML
TCIAA1059-25MLEA 386 DKK
TCIAA1059-25ML TCIAA1059-500ML
2-Hydroxyethyl acetate >60.0% (by GC)
2-Hydroxyethyl acetate
Formel: C₄H₈O₃
Kogepunkt: 182°C
Flammepunkt: 102°C
MDL Number: MFCD00002864
CAS-nummer: 542-59-6
EINECS: 208-821-8
Merck Index: 14,03801
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4522387733669dca71066f522993c45b04d201f5&vid=4522387733669dca71066f522993c45b04d201f5&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 60.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR