N-Boc-ethylenediamine ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
t-boc-N-EDA
A1371-1G
A1371-25G
A1371-5G
TCIAA1371-1GEA
156
DKK
TCIAA1371-1G
TCIAA1371-25G
TCIAA1371-5G
N-Boc-ethylenediamine ≥97.0% (by GC)
N-Boc-ethylenediamine
Formel:
C₇H₁₆N₂O₂ Kogepunkt: 105°C |
MDL Number:
MFCD00191871 CAS-nummer: 57260-73-8 UN nummer: 2735 ADR: 8,III |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...