Phenyl glycidyl ether ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Glycidyl phenyl ether
G0098-25G
G0098-500G
TCIAG0098-25GEA
132
DKK
TCIAG0098-25G
TCIAG0098-500G
Phenyl glycidyl ether ≥99.0% (by GC)
Phenyl glycidyl ether
Formel:
C₉H₁₀O₂ Molekylvægt: 150.18 g/mol Flammepunkt: 125°C |
MDL Number:
MFCD00005133 CAS-nummer: 122-60-1 EINECS: 204-557-2 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...