Phenyl glycidyl ether 99%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Glycidyl phenyl ether
135350500 135352500
ACRO135350500EA 483 DKK
ACRO135350500 ACRO135352500
Phenyl glycidyl ether 99%
Phenyl glycidyl ether
Formel: C₉H₁₀O₂
Molekylvægt: 150.18 g/mol
Kogepunkt: 245 °C (760 mmHg)
Smeltepunkt: 3.5 °C
Massefylde: 1.1 g/cm³
Flammepunkt: 115 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005133
CAS-nummer: 122-60-1
EINECS: 204-557-2
ADR: 6.1
Bestil nu
Specifikationer Testresultater
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5295 to 1.5315 (20°C, 589 nm)
GC ≥98.5 %