Ethyl-2-amino-4-methylthiazole-5-carboxylate 97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Ethyl 2-amino-4-methylthiazole-5-carboxylate
A16896.06
A16896.14
A16896.06EA
837
DKK
A16896.06
A16896.14
Ethyl-2-amino-4-methylthiazole-5-carboxylate 97%
Ethyl-2-amino-4-methylthiazole-5-carboxylate
Purity: 97%
CAS No.: 7210-76-6
Molecular Formula: C7H10N2O2S
Formula Weight: 186.22
Odour: Odourless
Melting Point: 172-176°
MDL No.: MFCD00123414
Formel:
C₇H₁₀N₂O₂S Molekylvægt: 186.22 g/mol Smeltepunkt: 172…176 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00123414 CAS-nummer: 7210-76-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...