2-Acetamido-5-nitrothiazole ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: N-(5-Nitro-2-thiazolyl)acetamide, N-(5-Nitrothiazol-2-yl)acetamide

A0689-1G A0689-5G
TCIAA0689-1GEA 403 DKK
TCIAA0689-1G TCIAA0689-5G
2-Acetamido-5-nitrothiazole ≥98.0% (by HPLC, titration analysis)
2-Acetamido-5-nitrothiazole
Formel: C₅H₅N₃O₃S
MDL Number: MFCD00022438
CAS-nummer: 140-40-9
EINECS: 205-414-7
Merck Index: 14,00410
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f927686d15c07f2abf67404adb5bfb654cb93645&vid=f927686d15c07f2abf67404adb5bfb654cb93645&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Amber powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Acetone within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR