3-Bromo-1-(trimethylsilyl)-1-propyne ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-(Trimethylsilyl)propargyl bromide, (3-Bromoprop-1-yn-1-yl)trimethylsilane
B1654-1G
B1654-5G
B1654-25G
TCIAB1654-1GEA
148
DKK
TCIAB1654-1G
TCIAB1654-5G
TCIAB1654-25G
3-Bromo-1-(trimethylsilyl)-1-propyne ≥97.0% (by GC)
3-Bromo-1-(trimethylsilyl)-1-propyne
Formel:
C₆H₁₁BrSi Molekylvægt: 191.14 g/mol Flammepunkt: 62°C |
MDL Number:
MFCD00134460 CAS-nummer: 38002-45-8 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...