Samarium diiodid ∼0.1 mol/L in THF

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

S0494-25ML S0494-100ML
TCIAS0494-25MLEA 427 DKK
TCIAS0494-25ML TCIAS0494-100ML
Samarium diiodid ∼0.1 mol/L in THF
Samarium diiodid
Formel: SmI₂
MDL Number: MFCD00058873
CAS-nummer: 32248-43-4
UN nummer: 2056
ADR: 3,II
Merck Index: 14,08349
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5f03330a2e6f1b2f65b304b6193016c93a52f21&vid=479327030d1961e96611f2bab01da4b391c885fc&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Blue to Dark blue clear liquid to cloudy liquid
Concentration (Titration) 0.075 to 0.105 mol/L


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR