9,10-Anthraquinone-2-carboxylic acid 99,0 Titrimetric

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Anthraquinone-2-carboxylic acid, 9,10-Dioxo-9,10-dihydroanthracene-2-carboxylic acid

A0504-5G A0504-25G
TCIAA0504-5GEA 600 DKK
TCIAA0504-5G TCIAA0504-25G
9,10-Anthraquinone-2-carboxylic acid 99,0 Titrimetric
9,10-Anthraquinone-2-carboxylic acid
Formel: C₁₅H₈O₄
Molekylvægt: 252.22 g/mol
MDL Number: MFCD00001231
CAS-nummer: 117-78-2
EINECS: 204-207-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ccdd8dfd339a284c87685ab0305dee0a668fe10f&vid=ccdd8dfd339a284c87685ab0305dee0a668fe10f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Amber powder to crystal
Purity (Neutralization titration) 99.0 to 101.0 %
Solubility in hot Acetic acid almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR