2-Amino-4-(4-pyridyl)thiazole
Leverandør: Cayman Chemical
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
4-(Pyridin-4-yl)thiazol-2-amine
11058-500
11058-1
11058-5
CAYM11058-500EA
1390
DKK
CAYM11058-500
CAYM11058-1
CAYM11058-5
2-Amino-4-(4-pyridyl)thiazole
2-Amino-4-(4-pyridyl)thiazole
Formel:
C₈H₇N₃S |
MDL Number:
MFCD00490009 CAS-nummer: 30235-28-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...