Iminostilbene 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 5H-Dibenzo[b,f]azepine

B21323.14 B21323.22 B21323.36
B21323.14EA 1090 DKK
B21323.14 B21323.22 B21323.36
Iminostilbene 97%
Iminostilbene

Purity: 97% CAS No.: 256-96-2 Molecular Formula: C14H11N Formula Weight: 193.25 Melting Point: 197-199° Beilstein Reference: 1343358

Formel: C₁₄H₁₁N
Smeltepunkt: 197…199 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005071
CAS-nummer: 256-96-2
EINECS: 205-970-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR