Zirkon(IV)oxid ≥99% (metals basis excluding Hf) stabilised, powder -325 Mesh (max. 4% Hf)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Zirconium dioxide, Zirconium Oxide
Stabilization: 10-15% Stabilised with yttria (Y₂O₃)
36272.22 36272.36
36272.22EA 1290 DKK
36272.22 36272.36
Zirkon(IV)oxid ≥99% (metals basis excluding Hf) stabilised, powder -325 Mesh (max. 4% Hf)
Zirkon(IV)oxid
Stabilized
Purity: 99% CAS No.: 1314-23-4 Molecular Formula: ZrO2 + 10-15% Y2O3 Physical Form: -325 Mesh Powder Appearance: White Odour: Odourless Boiling Point: ca 5000° Merck Reference: 14,10180 MDL No.: MFCD00145546
Formel: ZrO₂
Molekylvægt: 123.22 g/mol
Kogepunkt: ∼5000 °C
Smeltepunkt: ∼2700 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011310
CAS-nummer: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180