Banded earplugs, WaveBand®
Leverandør: Moldex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6800 6825
MOLD6800EA 6100 DKK
MOLD6800 111-1164
Banded earplugs, WaveBand®
Ørepropper Bøjlehøreværn
These anatomically designed banded earplugs are designed to be worn under the chin to allow freedom of movement without contact sounds.
  • Dual material technology reduces noise transmission
  • Soft anti-slip rubber grips allows easy on and off application
  • Extremely lightweight – Only 12 g
  • Band pressure only 1,2 N for high level of comfort
  • Replacement pods are available separately
Suitable for use against intermittent noise throughout the day.
SNR: 27 dB
Certificeringer: EN 352-2