Reusable half masks against gas, vapour and dust, 7000
Leverandør: Moldex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7001 7002 7003 7232 7432
111-0712EA 264 DKK
111-0712 111-0711 111-0710 111-1165 111-1168
Reusable half masks against gas, vapour and dust, 7000
Åndedrætsværn Genanvendelige masker Halvmasker
Reusable mask made from soft, durable material.
  • Adjustable head cradle
  • Easy maintenance and cleaning without tools
  • With EasyLock® filter system - easy handling
  • No additional adapters or parts necessary
  • Drop down option - mask can be worn around the neck during breaks
Certificeringer: EN 140:1998