Antibiotic assay discs
Leverandør: MACHEREY-NAGEL
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
484000
484001
MANA484000EA
432
DKK
MANA484000
MANA484001
Antibiotic assay discs
Mikrobiologiske testsystemer
Antibiotika test
These products are used in testing the resistance of pathogens to antibiotics. For this test the filter paper sections can be impregnated with the antibiotic to be tested and placed on the inoculated nutrient medium. Depending on the effectiveness, a smaller or larger zone of inhibition is formed.
- Non-impregnated filter paper sections
- Basis weight: 270 g/m²