Millicell® Cell Culture Inserts, PC Membrane (Isopore™)
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PIHP03050 PIXP01250 PI8P01250 PIHP01250 PITP01250
MILFPIHP03050EA 4180 DKK
MILFPIHP03050 MILFPIXP01250 MILFPI8P01250 MILFPIHP01250 MILFPITP01250
Millicell® Cell Culture Inserts, PC Membrane (Isopore™)
Mikroplader Indsatsplader
With Millicell inserts, attachment or suspension cells can access media from both their apical and basolateral sides. Cell growth, structure, and function more closely mimic what occurs in vivo. In addition, Millicell inserts make it possible to study both sides of the cell monolayer.
The inserts are easily prepared for SEM and TEM visualising techniques, and they are compatible with cellular and/or fluorescent stains.
Bestil nu
Specifikationer
Effektiv membrandiameter 30 mm 12 mm
Environmentally Preferable
Overall height 13 mm 10,5 mm